Tubing Extension Set, Arium®
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H2O-ATES-P H2O-ATES-UP
171-1397EA 178 EUR
171-1397 171-1398
Tubing Extension Set, Arium®
Waterdispensers
Extension tubing set (4 m) for the Arium® Smart Station is designed to provide uncomplicated extension of the connection between the Arium® Smart Station and the connected water system.
  • Flexible extension enhances the working space
  • Extend the operating range up to 4 meters
  • Fast and reliable connnection
The extended tubing set is used for the Smart Station Ultrapure and Pure for uncomplicated extension of the standard supplied tubing set. With the extension set the operating range can be easily increased from 2 to 4 meters.