b.safe Funnels 130
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R800-20 R801-40 R810-23 R811-40 R820-20 R820-23 R821-40 R830-20 R830-23 R831-40 R840-20 R840-23 R841-40 R850-20 R850-23 R851-40 R860-23 R861-40
590-0361EA 120 EUR
590-0361 590-0363 590-0365 590-0367 590-0369 590-0370 590-0372 590-0375 590-0376 590-0378 590-0383 590-0384 590-0388 590-0391 590-0392 590-0394 590-0396 590-0398
b.safe Funnels 130
Trechters Veiligheidstrechters
Funnel made of PE or static dissipative PE-EX for liquid waste disposal. The funnel can be safely mounted on the waste container with its freely turnable cap that is safely fixed with filling neck.
  • Smooth-running hinged lid protects from harmful vapours
  • Removable sieve restrains e.g. magnetic stirring bars or other solids
  • Small installation height ideal for integration into safety cabinets
  • All static dissipative funnels are supplied with assembled grounding cable (lengh: 1,5 m)
  • Free turnable cap for easy assembly on collecting container
  • Neck with plastic