Thread adaptors, TYPE B, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M600-02 M600-04 M615-01 M615-03 M615-11 M615-12
590-0233EA 141 EUR
590-0233 590-0234 590-0243 590-0244 590-0245 590-0246
Thread adaptors, TYPE B, b.safe
Fittingen en connectoren
All adaptors of this type are supplied with two male threads and recessed grips.
  • Stay flexible and independent
  • With recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.