Thread adaptors TYPE C, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M615-73 M615-80 M615-84 M615-87 M615-89 M630-83
590-0267EA 64.7 EUR
590-0267 590-0269 590-0270 590-0271 590-0272 590-0276
Thread adaptors TYPE C, b.safe
Fittingen en connectoren
All adaptors of this type are supplied with two female threads and recessed grips.
  • Stay flexible and independent
  • With recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.