Thread adaptors, TYPE D, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M520-01 M521-01
590-0206EA 94.9 EUR
590-0206 590-0207
Thread adaptors, TYPE D, b.safe
Adapters Adapters met NS-verbindingen
For assembly of components with ground joint cone NS 29/32 on bottles and vessels with GL 45 thread.
  • Stay flexible and independent
  • With recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.