Spouts, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R530-04 R530-05 R530-07 R530-09 R532-05 R532-07 R535-07
590-0322EA 22.7 EUR
590-0322 590-0323 590-0324 590-0325 590-0326 590-0327 590-0328
Spouts, b.safe
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
Screw cap for canister thread, freely turnable spout tube, rigid or flexible design with integrated safety aeration for constant flow.
  • Spout tube is freely turnable
  • Integrated safety aeration
  • Prevents loss of fluids
Quick refilling of liquids without dripping directly from canister.