Canisters, S90, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R265-10 R268-10
590-0303EA 32 EUR
590-0303 590-0304
Canisters, S90, b.safe
Veiligheidscontainers
Canister with thread S90 (DIN90) for the collection of liquid waste. Canister S90 in PE-EX with transparent stripe for observing the fill level.
  • Including screw cap
  • With UN-approval
  • b.safe accessories such as collecting trays, spouts and grounding cables, for dissipative canisters available
b.safe offers also suitable containers like flasks and canisters in addition to the wide range of caps for the extraction of solvents and waste caps or funnels for the collection of liquid waste.
Unless otherwise indicated, all b.safe canisters are supplied withMore Product Information