Canisters, S60/61, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R230-05 R230-10 R230-20 R231-10 R231-30 R236-10 R236-20
590-0293EA 24.7 EUR
590-0293 590-0294 590-0295 590-0296 590-0297 590-0298 590-0299
Canisters, S60/61, b.safe
Veiligheidscontainers
Canister with S60/61 thread (DIN60/61) for the collection of liquid waste.
  • Including screw cap
  • With UN-approval
  • b.safe accessories such as collecting trays, spouts and grounding cables, for dissipative canisters available
b.safe offers also suitable containers like flasks and canisters in addition to the wide range of caps for the extraction of solvents and waste caps or funnels for the collection of liquid waste.
Unless otherwise indicated, all b.safe canisters are supplied with UN-approval and can be used for the transport of hazardous goods.