Canisters, S55, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R225-05 R225-10
590-0291EA 23.5 EUR
590-0291 590-0292
Canisters, S55, b.safe
Veiligheidscontainers
Canister with S55 thread (DIN51) for the collection of liquid waste.
  • Including screw cap
  • With UN-approval
  • b.safe accessories such as collecting trays, spouts and grounding cables, for dissipative canisters available
b.safe offers also suitable containers like flasks and canisters in addition to the wide range of caps for the extraction of solvents and waste caps or funnels for the collection of liquid waste.
Unless otherwise indicated, all b.safe canisters are supplied with UN-approval and can be used for the transport of hazardous goods.