Canisters, S60/61, with level indicator, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R237-10 R237-20
590-0300EA 122 EUR
590-0300 590-0301
Canisters, S60/61, with level indicator, b.safe
Veiligheidscontainers
Canister with S60/61 thread (DIN 60/61) for the collection of liquid waste with integrated indicator to monitor the level.
  • Including screw cap
  • With level indicator
  • b.safe accessories such as collecting trays, spouts and grounding cables, for dissipative canisters available
The red visual pin goes up with increasing filling level. As soon as the visual pin fills the inspection glass completely, the canister is full and has to be exchanged Unless otherwise indicated, all b.safe canisters are supplied with UN-approval and can be used for the transport of hazardous goods.
Order Now
Specificaties
Environmentally Preferable