Politainers, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R295-05 R295-10 R297-20
590-0305EA 10.7 EUR
590-0305 590-0306 590-0307
Politainers, b.safe
Veiligheidscontainers
Foldable canister for the collection of liquid waste.
  • Including screw cap
  • Foldable canister
Folded to save space during delivery and storage, unfolds automatically during filling. UN-approval only if used with the appropriate carton box.
Bestelinformatie: Please order additional accessories like the carton box or the adaptor for Waste Caps GL 45 separately.