Capillary cutter, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R570-01 R572-05 R573-05
590-0334EA 70.9 EUR
590-0334 590-0335 590-0336
Capillary cutter, b.safe
Snijders
Ideal for cutting capillaries and elastic tubes without reinforcement.
  • Capillary cutter for up to tubing Ext.Ø 8 mm with guide bores
  • High qualitiy capillary cutter
  • Replacement blades available
The guide bores help to get a clean cut of the capillaries.
Order Now
Specificaties
Environmentally Preferable