Label maker, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M550-01 M550-02
590-0217EA 164 EUR
590-0217 590-0218
Label maker, b.safe
Printers Label Printers
Portable label printer with computer-style keyboard, large LCD display and 20 methods to format text.
  • Portable
  • Powered batteries or optional AC-adapter
Powered by 6 batteries AAA or optional AC-adapter (both not included).
Leveringsinformatie: Includes one label cassette black on white, width 12 mm/length 3 m. Labels up to a width of 12 mm can be printed.