Caps for solvent supplies, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M128-01 M128-02 M138-01 M138-02 M138-03 M139-01 M139-02 M139-03 M140-01 M140-02 M140-03 M141-01 M143-01 M143-02 M143-03 M144-01 M144-02 M144-03 M144-05 M145-01 M145-02 M145-03 M145-04 M145-05 M147-01 M147-02 M180-01 M180-02 M180-03 M183-04 M529-01
590-0063EA 160 EUR
590-0063 590-0064 590-0065 590-0066 590-0067 590-0068 590-0069 590-0070 590-0071 590-0072 590-0073 590-0074 590-0075 590-0076 590-0077 590-0078 590-0079 590-0080 590-0081 590-0082 590-0083 590-0084 590-0085 590-0086 590-0090 590-0091 590-0092 590-0093 590-0094 590-0097 590-0208
Caps for solvent supplies, b.safe
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
b.safe caps together with fittings, tubing and an air valve form a perfect unit for solvent extraction from laboratory flasks.
  • Perfect protection from hazardous vapours
  • Closed system: reduction of expensive solvent loss
  • Guarantee of pure eluents
  • Huge choice of cap sizes for compatibility with any eluent flask
  • No twisting of capillaries
  • Screw caps can individually be labelled by hand or with removable, self-adhesive labels
b.safe air valve: The special valve lets flow into the bottle as much ambient air as necessary for pressure compensation duringMore Product Information