Blind fittings, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M501-01 M501-45 M501-50 M501-55
590-0147EA 49.3 EUR
590-0147 590-0155 590-0156 590-0157
Blind fittings, b.safe
Fittingen en connectoren
Made of PFA to close unused connections (thread UNF ¹/₄" 28G) on b.safe caps and b.safe waste caps.
  • Internal cone for comfortable inlet of capillaries
  • Optimized surface design for more grip
  • Centring pin for easy installation
For tight closure of unused connectors on b.safe caps and b.safe waste caps. With ergonomic gripping surface or knurl for easy assembly. The version with ground joint has an integrated nut to loosen stuck ground joints.