Hose connectors, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M503-01 M503-05 M503-08 M503-09 M503-10 M503-11 M503-12 M503-13 M503-14 M503-15 M503-16 M513-01 M513-02 M518-01 M519-01
590-0162EA 14.5 EUR
590-0162 590-0163 590-0164 590-0165 590-0166 590-0167 590-0168 590-0169 590-0170 590-0171 590-0172 590-0197 590-0198 590-0204 590-0205
Hose connectors, b.safe
Fittingen en connectoren
For the connection of elastic tubing (e.g. Viton, Tygon, silicone) on b.safe waste caps.
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
b.safe hose connectors are the flexible choice to customise your b.safe waste cap. Made of PP or PTFE and available in numerous diameters you will find the exact one to fit your waste hose.
Order Now
Specificaties
Environmentally Preferable