Tubing connectors, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M590-03 M590-05 M590-07
590-0228EA 9.4 EUR
590-0228 590-0229 590-0230
Tubing connectors, b.safe
Fittingen en connectoren
Straight fitting made of PP, with two hose connectors for elastic tubing.
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
b.safe hose connectors are the flexible choice to customise your b.safe waste cap. Made of PP or PTFE and available in numerous diameters you will find the exact one to fit your waste hose.