Corrugated tubing couplings, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M514-03 M514-05
590-0199EA 55.8 EUR
590-0199 590-0200
Corrugated tubing couplings, b.safe
Fittingen en connectoren
Three-part coupling consisting of coupling, locking nut made of PP and clamp ring made of FEP. To connect hard-walled corrugated tubing with the hose connection (NPT ¹/₈" thread) on b.safe waste caps. Locking nut with ergonomic gripping surface, additional grip hollows with SW10 on the coupling.
  • Alternative to hose connectors
  • Especially for corrugated hoses
  • With locking nut and clamp ring
  • Reliably seals the flexible corrugated tubing of your HPLC
Choose the b.safe corrugated tubing coupling as per your HPLC tubing. Devices of Agilent Technologies for example contain corrugated tubing with int. Ø of 6,5 mm. Devices of Thermo Scientific contain corrugated tubing with int. Ø of 8,5 mm.