Tubing connectors Y, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M580-02 M580-04 M580-06 M580-08 M580-10 M580-12
590-0222EA 12 EUR
590-0222 590-0223 590-0224 590-0225 590-0226 590-0227
Tubing connectors Y, b.safe
Fittingen en connectoren
Y-shaped fitting made of PP, with three hose connectors for elastic tubing.
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
b.safe hose connectors are the flexible choice to customise your b.safe waste cap. Made of PP or PTFE and available in numerous diameters you will find the exact one to fit your waste hose.