Multiports, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M509-34 M509-36 M509-25 M509-26 M509-28 M509-29 M509-32 M509-33
BOHLM509-34EA 66 EUR
BOHLM509-34 BOHLM509-36 590-0183 590-0184 590-0185 590-0186 590-0187 590-0188
Multiports, b.safe
Fittingen en connectoren
Extension of the number of capillary and/or tubing connectors on a b.safe Waste Cap. With PFA Fittings for capillaries with Ext.Ø 1,6/2,2/3,2 mm (thread UNF ¹/₄"), 1 hose connector for tubing Int.Ø 5,5 to 8,0 mm (thread NPT ¹/₈").
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary and tubing connectors of a b.safe waste cap.