Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BIMI2138-100MG BIMI2138-10MG BIMI2138-50MG
APOSBIMI2138-100MGEA 454 EUR
APOSBIMI2138-100MG APOSBIMI2138-10MG APOSBIMI2138-50MG
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Eiwitten en peptiden

  • Free base
  • GHS symbol and signal word: GHS07 and warning
  • Melting point: 245 °C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR