NANOCOLOR® Photometric Tube Test Kits for Copper
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
985054 985053
MANA985054EA 99.8 EUR
MANA985054 MANA985053
NANOCOLOR® Photometric Tube Test Kits for Copper
Milieutestkits
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of the copper in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.
Sea water analysis: Yes
Hazardous material: No
Remark: Measuring range on NANOCOLOR VIS II
Leveringsinformatie: Rugged box with 20 test tubes.