Coronavirus and SARS-CoV-2 direct RT real-time PCR kits, GSD NovaPrime®
Leverancier: GOLD STANDARD DIAGNOSTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PCOV6043 PCOV6044
GSDCPCOV6043EA 806 EUR
GSDCPCOV6043 GSDCPCOV6044
Coronavirus and SARS-CoV-2 direct RT real-time PCR kits, GSD NovaPrime®
Diagnostische systemen Clinical Reagents and Kits
This Real-Time RT-PCR assay is intended for the qualitative detection of SARS-CoV-2 from human nasopharyngeal swabs, eluted directly into deionised water without an additional RNA extraction process.
  • Simplify the process for COVID-19 testing
  • Meet the increasing global demand for molecular solutions
  • Maximize accessibility of RT-PCR testing to a larger number of laboratories
  • Increase test processing
  • Reduce the cost, labour, and time required to perform RT-PCR
This new workflow shortens the complete turnaround time of the test, providing results in less than 2 hours from sample to result without compromising the high accuracy of PCR testing.
Certificaten: CE-IVD marked.