Coronavirus and SARS-CoV-2 ELISA kits, NovaLisa®
Leverancier: GOLD STANDARD DIAGNOSTICSTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
This ELISA kit is intended for the qualitative detection of IgA class antibodies to SARS-CoV-2 virus in human serum and plasma for diagnostics of exposure to coronavirus (COVID-19).
- Separate detection of IgG, IgM and IgA antibodies
- Advantages of the NovaTec reagent system
- Total incubation under 2 hours
Compatible with diverse open platform ELISA analysers.
Certificaten: CE-IVD marked.