Benzyl 2-acetamido-6-O-benzyl-2-deoxy-4-O-beta-D-galactopyranosyl-alpha-D-glucopyranoside ≥97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BICL5022-25MG BICL5022-50MG BICL5022-100MG
APOSBICL5022-25MGEA 338 EUR
APOSBICL5022-25MG APOSBICL5022-50MG APOSBICL5022-100MG
Benzyl 2-acetamido-6-O-benzyl-2-deoxy-4-O-beta-D-galactopyranosyl-alpha-D-glucopyranoside ≥97%
Benzyl 2-acetamido-6-O-benzyl-2-deoxy-4-O-beta-D-galactopyranosyl-alpha-D-glucopyranoside