1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)cytosine hydrochloride (2’-FANA-C) ≥97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIMN2025-1G BIMN2025-250MG BIMN2025-500MG
APOSBIMN2025-1GEA 1360 EUR
APOSBIMN2025-1G APOSBIMN2025-250MG APOSBIMN2025-500MG
1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)cytosine hydrochloride (2’-FANA-C) ≥97%
1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)cytosine hydrochloride (2’-FANA-C)