4-Bromo-2-fluoro-1-(4-methoxyphenoxy)benzene ≥95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC52475-10G PC52475-1G PC52475-5G
APOSPC52475-10GEA 577 EUR
APOSPC52475-10G APOSPC52475-1G APOSPC52475-5G
4-Bromo-2-fluoro-1-(4-methoxyphenoxy)benzene ≥95%
4-Bromo-2-fluoro-1-(4-methoxyphenoxy)benzene