Nitrate CRM, standard solutions, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.25036.0100 1.25037.0100 1.25038.0100 1.25039.0100 1.25040.0100 1.32240.0100 1.32241.0100 1.32242.0100
1.25036.0100EA 51.6 EUR
1.25036.0100 1.25037.0100 1.25038.0100 1.25039.0100 1.25040.0100 1.32240.0100 1.32241.0100 1.32242.0100
Nitrate CRM, standard solutions, Supelco®
Standaarden Anionische standaarden Nitraat standaarden
NaNO₃ in H₂O
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Certificaten: Traceable to SRM from NIST and USP.