Silicate CRM, Standard Solutions, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.32243.0100 1.32244.0100 1.32245.0100
1.32243.0100EA 60.5 EUR
1.32243.0100 1.32244.0100 1.32245.0100
Silicate CRM, Standard Solutions, Supelco®
Standaarden Anionische standaarden Silicaat standaarden
SiO₂ in H₂O
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Certificaten: Traceable to SRM from NIST and USP.