Coronavirus and SARS-CoV-2 Ag rapid test kits, GSD NovaGen
Leverancier: NovaTec Immundiagnostics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GSD NovaGen
GSDCCVAG4080BEA 91.7 EUR
GSDCCVAG4080B GSDCCVAG4080A
Coronavirus and SARS-CoV-2 Ag rapid test kits, GSD NovaGen
Diagnostische systemen Clinical Reagents and Kits
GSD NovaGen SARS-CoV-2 Ag rapid test completes the diagnostic panel of the Gold Standard Diagnostics Group for SARS-CoV-2 diagnostics: ELISA, RT-PCR, and RNA extraction kits. This lateral-flow test meets the WHO performance criteria (sensitivity higher than 80%; specificity higher than 97%) for use in the global fight against COVID-19.
  • Current mutant strains can be detected (UK, South Africa, Brazil)
  • Easy specimen collection: nasal and nasopharyngeal swabs
  • Fast results in 15 minutes
  • Intuitive visual interpretation
  • High sensitivity and specificity
  • Easy to perform: no special equipment needed
It is intended for the qualitative detection of SARS-CoV-2 Nucleocapsid (N) Protein antigens present in respiratory samples.
Certificaten: CE-IVD marked.