Cathepsin/subtilisin inhibitor
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIC2003-1MG
BIC2003-5MG
APOSBIC2003-1MGEA
162
EUR
APOSBIC2003-1MG
APOSBIC2003-5MG
Cathepsin/subtilisin inhibitor
Eiwitten en peptiden
Selectively inhibits members of Cystein protease family including Cathepsin L and members of Serine protease family including Subtilisin, Carlsberg and Thermitase