Phosphoramidon disodium salt
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIMI0549-10MG BIMI0549-5MG
APOSBIMI0549-10MGEA 285 EUR
APOSBIMI0549-10MG APOSBIMI0549-5MG
Phosphoramidon disodium salt
Eiwitten en peptiden
Inhibitor of thermolysin. Weak collagenase inhibitor. Does not inhibit chymotrypsin, papain, pepsin or trypsin.