Soybean trypsin inhibitor
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIMI1441-1G BIMI1441-250MG BIMI1441-5G
APOSBIMI1441-1GEA 213 EUR
APOSBIMI1441-1G APOSBIMI1441-250MG APOSBIMI1441-5G
Soybean trypsin inhibitor
Eiwitten en peptiden
Assay >10000 U/mg lyophilizedInhibitor of Trypsin, Chymotrypsin, Plasmin and otherproteolytic enzymes.