Leupeptin hemisulphate
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIMI2183-100MG BIMI2183-25MG BIMI2183-50MG
APOSBIMI2183-100MGEA 301 EUR
APOSBIMI2183-100MG APOSBIMI2183-25MG APOSBIMI2183-50MG
Leupeptin hemisulphate
Eiwitten en peptiden
Leupeptin is a peptide antibiotic, isolated from several Streptomyces species. It is a reversible inhibitor ofserine and cysteine proteases