Leupeptin hydrochloride
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIMI2442-100MG BIMI2442-250MG BIMI2442-25MG
APOSBIMI2442-100MGEA 220 EUR
APOSBIMI2442-100MG APOSBIMI2442-250MG APOSBIMI2442-25MG
Leupeptin hydrochloride
Eiwitten en peptiden
Inhibitor of serine and cysteine proteases
Order Now
Specificaties
Protein/peptide name Leupeptin hydrochloride
Environmentally Preferable
Protein synonyms Acetyl-Leu-Leu-Arg-al hydrochloride
Species
Bron
Zuiverheid >90%