H-FLEX HS4 windshield swinging bucket rotor and accessories
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75003330 75003338 75003339
521-3440EA 4100 EUR
521-3440 521-3441 521-3442
H-FLEX HS4 windshield swinging bucket rotor and accessories
Centrifuge Vloercentrifuges
Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor at 7197xg in 4 L High Performance General Purpose Centrifuges.
  • SMARTSpin™ technology
  • Space saving design
Swing bucket rotor enables fast Auto-Lock, one-handed rotor exchanges. Spins blood tubes, conical tubes and microplates without changing rotor or buckets. Results are faster without the need to change hardware. Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants.
Swing bucket rotor can be used with equipments such as Sorvall X4 Pro, Sorvall X4 F Pro, Multifuge X4 Pro, Multifuge X4 F Pro, Sorvall ST4 Plus, Sorvall ST4F Plus, Megafuge ST4 Plus, Megafuge ST4F Plus.