Freestanding centrifuge tubes, 50 ml, PerformR®
Leverancier: LABCON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3094-340-008-9 3094-345-008-9 3094-870-008-9 3095-345-008-9 3095-870-008-9
3094-340-008-9EA 394 EUR
3094-340-008-9 3094-345-008-9 3094-870-008-9 3095-345-008-9 3095-870-008-9
Freestanding centrifuge tubes, 50 ml, PerformR®
Buisjes Centrifugebuisjes
PerformR® freestanding centrifuge tubes provide reliable performance in major centrifuges. With spin rates higher than most popular brands, these tubes are made from high-quality lab certified non-cytotoxic medical grade resin.
  • ATP-, RNase/DNase-, and endotoxin-free (non-pyrogenic)
  • Certified low trace metals (1 ppm)
  • White writing surface
  • Polypropylene USP Class VI resin
  • Free of DiHEMDA, oleamide, and BPA
  • Freezable to –80 °C, and autoclavable
  • Validated Sterilisation (ISO 11137) for use in Research and Medical labs
PerformR® freestanding centrifuge tubes are available with an easily labelled flat cap, or a deep plug seal cap for a secure seal when using shakers and horizontal incubators.
Verpakking: Bulk packed items are supplied with tubes and caps in separate bags. Bagged tubes include 50 per bag, 10 bags per case.