Polypropylene centrifuge tubes, PerformR®
Leverancier: LABCON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3136-345-008-9 3137-345-008-9 3146-335-018-9 3146-870-008-9 3147-335-018-9 3147-870-008-9 3186-345-008-9 3187-345-008-9 3196-335-018-9 3196-870-008-9 3197-335-018-9 3197-870-008-9
3136-345-008-9EA 299 EUR
3136-345-008-9 3137-345-008-9 3146-335-018-9 3146-870-008-9 3147-335-018-9 3147-870-008-9 3186-345-008-9 3187-345-008-9 3196-335-018-9 3196-870-008-9 3197-335-018-9 3197-870-008-9
Polypropylene centrifuge tubes, PerformR®
Buisjes Centrifugebuisjes
PerformR® centrifuge tubes combine reliable performance in major centrifuges with spin rates higher than many popular brands. The high-quality, lab certified, non-cytotoxic medical grade resin means users don't have to compromise their work with low-quality tubes. 
  • ATP-, RNase/Dnase-, and endotoxin-free (non-pyrogenic)
  • Black printed graduations and white writing spot
  • Validated Sterilisation (ISO 11137) for use in Research and Medical labs
  • Polypropylene USP Class VI resin
  • Freezable to –80 °C, and autoclavable
They are designed for everyday centrifugation and storage needs. These tubes include large white solvent-resistant labeling areas and black easy-to-read graduations. They are available with either a flat cap that is easy to label, or a plug style cap for horizontal applications.
Racked tubes are packaged in a reusable, autoclavable, and freezable (–80 °C) polypropylene rack that inter-locks to allow you to create a workstation.
Tubes can be ordered with a flat cap that can easily be labeled or a plug style cap that includes a very deep sealing area. The flat caps are designed to allow an evaporation seal with ¾ turn for one-handed operation. And then a final ¼ turn will seal them liquid tight.
Verpakking: Bulk packed items are supplied with tubes and caps in separate bags. Bagged tubes include 50 per bag, 10 bags per case. Reusable polypropylene rack includes 25 tubes per rack, 2 racks per pack, 20 packs per case.