TE Buffers 100X
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R003 786-752 786-813
R003EA 95.8 EUR
R003 786-752 786-813
TE Buffers 100X
Buffers Biologische buffers
TE Buffer (Tris-EDTA Buffer) is a common molecular biology buffer that is used to protect DNA and RNA from degradation. The Tris buffering agent and EDTA metal chelating properties help protect DNA and RNA.