Accessories for PLATEMASTER®
Leverancier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F1077602 F1077605 F1077606 F1077604 F1077603 F110767
613-5910EA 1664 EUR
613-5910 631-1373 631-1374 631-1372 631-1359 GILSF110767
Accessories for PLATEMASTER®
Automated Liquid Handling Systems
The Pipetting Head Height Adjuster is an accessory for the PLATEMASTER® that allows for easy altering of pipetting height, ensuring appropriate tip depth within shallow- or deep-well microplates.
The Lubrication Box of O-rings is an accessory for the PLATEMASTER®. We recommend lightly lubricating the external O-ring between regular maintenance intervals in situations of intensive use and to avoid ill-fitting tips.