GC columns J&W CP-Wax 52 CB
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7178 CP8543I CP7743I CP7698I CP7791I CP7658II5 CP8713II5 CP7773II5 CP8073II5 CP7723II5 CP7673II5 CP7773I CP7668I CP8718I
VARICP7178EA 856 EUR
VARICP7178 VARICP8543I VARICP7743I VARICP7698I VARICP7791I VARICP7658II5 VARICP8713II5 VARICP7773II5 VARICP8073II5 VARICP7723II5 VARICP7673II5 VARICP7773I VARICP7668I VARICP8718I
GC columns J&W CP-Wax 52 CB
Kolommen voor chromatografie
CP-Wax 52 provides higher resolution of low boiling point analytes, and is ideal for a variety of EPA and ASTM methods.
  • Polyethylene glycol (PEG) phase
  • High polarity- provides separations for a broad range of applications
  • Excellent column-to-column inertness reproducibility
  • Sharp and symmetrical peak shape performance for active polar compounds
  • Bonded and cross-linked; solvent rinsable
CP-Wax 52 CB GC columns have lower minimum temperature and a higher maximum temperature than non-bonded polyethylene glycol (PEG) columns due to extensive cross linking, delivering higher resolution of low-boiling-point analytes. With guaranteed reproducibility and excellent temperature stability, CP-Wax 52 CB is ideal for EPA and ASTM methods.
The improved line of Agilent J&W CP-Wax 52 CB GC columns utilises the same innovative inert processes employed in the manufacture of Agilent J&W DB-WAX Ultra Inert columns. These columns have an extended inertness lifetime that withstands repeated temperature cycling to the upper temperature limits of the column, while critical performance parameters such as selectivity, theoretical plates and retention indices remain unchanged.