HyClone CDM4PERMAb
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30871.02 SH30872.01
SH30871.02EA 95.9 EUR
SH30871.02 HYCLSH30872.01
HyClone CDM4PERMAb
Celcultuurmedia
HyClone™ CDM4PERMAb is a high-performance cell culture medium developed to increase process yields in the production of human antibodies and recombinant proteins using PER.C6 (Crucell NV) technology. Manufactured according to cGMP guidelines.
  • Animal-derived component free (ADCF)
  • Chemically-defined formulation
  • Metabolically designed for high cell yield and recombinant protein production
  • Allows for direct or sequential adaptation