Fittings for Shimadzu LC systems
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8001-0803 8001-0805 8001-0806 8001-0813 8001-0816 8001-0817 8001-0819
HEWL8001-0803EA 0 EUR
HEWL8001-0803 HEWL8001-0805 HEWL8001-0806 HEWL8001-0813 HEWL8001-0816 HEWL8001-0817 HEWL8001-0819
Fittings for Shimadzu LC systems
Fittingen en connectoren
These fittings for Shimadzu HPLC systems provide leak-free connections and prevent the loss of peak shape and resolution.
  • Robust design for long life and smooth, leak-free operation
  • Stainless steel nuts and ferrules for high-pressure systems
To ensure leak-free connections, and to prevent the loss of peak shape and resolution, always use the recommended fitting style for columns, valves, and unions.