GC columns, Ultra Inert, J&W DB-WAX
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
122-7033UI 122-7012UI 122-7062UI 121-7022UI 121-7023UI 122-7063UI 123-7012UI 123-7032UI 123-7033UI 123-7062UI 123-7063UI 125-7012UI 125-7031UI 125-7032UI 125-7037UI 125-7062UI 128-7022UI
HEWL122-7033UIEA 803 EUR
HEWL122-7033UI HEWL122-7012UI HEWL122-7062UI HEWL121-7022UI HEWL121-7023UI HEWL122-7063UI HEWL123-7012UI HEWL123-7032UI HEWL123-7033UI HEWL123-7062UI HEWL123-7063UI HEWL125-7012UI HEWL125-7031UI HEWL125-7032UI HEWL125-7037UI HEWL125-7062UI HEWL128-7022UI
GC columns, Ultra Inert, J&W DB-WAX
Kolommen voor chromatografie
GC columns achieve the highest levels of PEG stationary phase inertness performance for a range of challenging active analytes with individually-tested DB-WAX UI columns.
  • Improved peakshape for challenging polar compounds with DB-WAX Ultra Inert
  • Reliable column to column inertness performance
  • Equivalent bonded polyethylene glycol (PEG) selectivity as DB-WAX; no method revalidation needed
  • High polarity, equivalent to USP G16
  • Excellent performance for acidic compounds including free fatty acids no need for FFAP type column
DB-WAX Ultra Inert (UI)More Product Information
Order Now
Specificaties
Environmentally Preferable