J&W HP-PLOT Al₂O₃ KCl GC columns
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19091P-K15 19091P-K15E 19091P-K33 19095P-K23 19095P-K25 19091P-K33LTM 19095P-K23LTM
HEWL19091P-K15EA 1504 EUR
HEWL19091P-K15 HEWL19091P-K15E HEWL19091P-K33 HEWL19095P-K23 HEWL19095P-K25 HEWL19091P-K33LTM HEWL19095P-K23LTM
J&W HP-PLOT Al₂O₃ KCl GC columns
Kolommen voor chromatografie
Agilent J&W HP-PLOT Al₂O₃ KCl is the least polar alumina phase and is deactivated with sodium chloride.
  • Least polar alumina phase
  • Low retention of olefins relative to comparable paraffin
  • Recommended phase for many ASTM methods
It is the standard column choice for light hydrocarbon analysis, particularly C1 to C8 hydrocarbon isomers. Excellent for the quantification of dienes, especially propadiene and butadiene from ethylene and propylene streams.