PFA inert sample introduction kit supplies
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G3285-80020 G3285-80021 G3285-80024 G4912-68000 G4912-68001 G4912-68002 G4912-68003 G4912-80010 G4912-80011 G4912-80012 G4912-80014 G4912-80016 G3285-80085
HEWLG3285-80020EA 714 EUR
HEWLG3285-80020 HEWLG3285-80021 HEWLG3285-80024 HEWLG4912-68000 HEWLG4912-68001 HEWLG4912-68002 HEWLG4912-68003 HEWLG4912-80010 HEWLG4912-80011 HEWLG4912-80012 HEWLG4912-80014 HEWLG4912-80016 HEWLG3285-80085
PFA inert sample introduction kit supplies
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Inert kit for ICP-MS enables analysis of high-purity and HF-containing samples using a PFA-based sample intro system with inert Pt or sapphire-injector torch.
  • The O-ring free design and PFA construction material ensure low Background Equivalent Concentrations (BEC) and Detection Limits (DL)
  • Added flexibility for organic solvent analysis with an optional connecting pipe with gas port for the addition of O₂/Ar
  • Compatible with PFA MicroFlow concentric nebulizer, PFA MicroFlow nebuliser with I-AS probe, PFA concentric 200 μl/min nebulizer and PFA concentric 200 µl/min nebulizer with probe
They manufacture a range of high quality ICP-MS replacements, accessories and consumables including PFA Inert Sample Introduction Kit Supplies. A full range of accessories are available including connector and drain tubes, injector assemblies and torches for 7500 and 7700 ICP-MS systems. To maintain system performance, ensure that consumables are periodically replaced in accordance with recommended maintenance schedules.
A complete kit containing a PFA end cap, PFA spray chamber, connecting tube, demountable quartz torch, torch injector (four options are available: platinum or sapphire with a 2,5 mm or 1,5 mm injector) with PFA base. Easy to install in the standard Peltier-cooled spray chamber mount of the 7700. System uses a simple push-fit torch connector to aid assembly and disconnection