Electron capture detectors (ECD)
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18713-60040 18713-60050 19233-20625 19233-20755 G1533-80565 G2397-20540 G2397-80520
HEWL18713-60040EA 91 EUR
HEWL18713-60040 HEWL18713-60050 HEWL19233-20625 HEWL19233-20755 HEWLG1533-80565 HEWLG2397-20540 HEWLG2397-80520
Electron capture detectors (ECD)
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
  • Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.