J.T.Baker®, Robotic Tips Type ST
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
49000-0000EA 1860 EUR
49000-0000 49000-0006 49001-0000 49001-0106 49000-1106 49001-1106 49000-3106 49001-3106 49000-2000
J.T.Baker®, Robotic Tips Type ST
Pipettips
Suitable for use on Euroimmun Analyzer. Available in conductive version, with or without filter, and in CE-IVD marked version for clinical diagnostic applications.
  • Manufactured under cleanroom conditions (cleanroom Class 8)
  • Conductive tips for liquid level sensing
  • Ideal for dosing even small volumes with high precision and without contamination
J.T.Baker® robotic tips type ST are suitable for use on Euroimmun Analyzer and other robotic instruments using the same pipetting head.
The validation of the tips on the instrument for specific applications isMore Product Information
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)