Method development kits
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5183-4624 5183-4625 5183-4626 5183-4627 5185-5807 5185-5808 5185-5809 5185-5810 5188-5323 5188-5324 5188-5328 5190-1432 5190-1436 5190-1435 5190-1433 5190-1431 5190-1434
HEWL5183-4624EA 1388 EUR
HEWL5183-4624 HEWL5183-4625 HEWL5183-4626 HEWL5183-4627 HEWL5185-5807 HEWL5185-5808 HEWL5185-5809 HEWL5185-5810 HEWL5188-5323 HEWL5188-5324 HEWL5188-5328 HEWL5190-1432 HEWL5190-1436 HEWL5190-1435 HEWL5190-1433 HEWL5190-1431 HEWL5190-1434
Method development kits
Kolommen voor chromatografie
Agilent offers methods development kits to streamline the column collection process used for medium and high pressure LC systems.
  • Kits based on changing pH or selectivity for methods development
  • Column selection options include Eclipse Plus C18, Eclipse Plus Phenyl-Hexyl, Bonus-RP, StableBond C18, and Extend-C18
  • Column size options include 2,1×50 mm, 4,6×50 mm, and 4,6×100 mm with 1,8 µm or 3,5 µm particle sizes
Method development is some of the most important work. Now, it's easier than ever to evaluate the best chemistries for separation with Agilent Method Development Kits. These kits include a range of column chemistries and selectivities, giving the tools needed to save time and perfect your separation. Agilent’s columns are manufactured to the industry's tightest quality control specifications for reliable, reproducible performance. We also offer the industry's widest range of Fast LC columns to get high resolution and speedy analysis times. Request the Method Development Poster! When done with method development, Agilent Method Validation Kits provide columns from three lots to validate the method.