HPLC columns, InfinityLab poroshell 120, phenyl-hexyl
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
695775-912 693975-912 699975-312 693975-312 699975-912 699775-912 695975-912 695975-312 823750-912 823750-914 821725-912 821725-914 820750-914 820750-912 699970-312 699970-912 695770-912 695970-312 695970-912 699770-912
HEWL695775-912EA 785 EUR
HEWL695775-912 HEWL693975-912 HEWL699975-312 HEWL693975-312 HEWL699975-912 HEWL699775-912 HEWL695975-912 HEWL695975-312 HEWL823750-912 HEWL823750-914 HEWL821725-912 HEWL821725-914 HEWL820750-914 HEWL820750-912 HEWL699970-312 HEWL699970-912 HEWL695770-912 HEWL695970-312 HEWL695970-912 HEWL699770-912
HPLC columns, InfinityLab poroshell 120, phenyl-hexyl
Kolommen voor chromatografie
Rugged, high-resolution separations up to five times faster than traditional 5 µm methods.
  • High efficiency columns for improved throughput
  • Excellent lot-to-lot reproducibility gives confidence in separation results
InfinityLab Poroshell 120 columns provide exceptional efficiency and reliability, significantly boosting performance from all instruments, whether need improved HPLC performance, UHPLC performance at lower pressures or the highest UHPLC performance. The three particle diameters 1,9 μm, 2,7 μm, and 4 μm allow to choose the size that best fits separation needs and is most compatible with LC systems, and 12 different chemistries provide a range of selectivities that makes your method development fast and easy.